Learn More
Cayman Chemical Desethoxy Quetiapine (hydrochloride), Purity: ≥95%, CAS: 329218-14-6, 50 mg
SDP
Catalog No. 502914005
Encompass_Preferred
An active metabolite of quetiapine; binds to dopamine D2 receptors (IC50 = 1,330 nM); reduces apomorphine-induced climbing in mice; reduces apomorphine-induced swimming deficits in mice
Catalog No. 50-291-4005 Supplier Cayman Chemical Supplier No. 3541050 MG
An active metabolite of quetiapine; binds to dopamine D2 receptors (IC50 = 1,330 nM); reduces apomorphine-induced climbing in mice; reduces apomorphine-induced swimming deficits in mice