Learn More
Cayman Chemical Desethyl Hydroxychloroquine-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1854126-47-8, 5 mg
SDP
Catalog No. 502990353
Encompass_Preferred
An internal standard for the quantification of desethyl hydroxychloroquine by GC- or LC-MS
Catalog No. 50-299-0353 Supplier Cayman Chemical Supplier No. 311625 MG
An internal standard for the quantification of desethyl hydroxychloroquine by GC- or LC-MS