Learn More
Cayman Chemical Desethylene Ciprofloxacin (hydrochloride), Purity: ≥98%, CAS: 528851-31-2, 10 mg
SDP
Catalog No. 502987306
Encompass_Preferred
A major metabolite and degradation product of ciprofloxacin; can be created through advanced oxygenation processes of ciprofloxacin in wastewater treatment; can be created by chlorination of ciprofloxacin
Catalog No. 50-298-7306 Supplier Cayman Chemical Supplier No. 2212310 MG
A major metabolite and degradation product of ciprofloxacin; can be created through advanced oxygenation processes of ciprofloxacin in wastewater treatment; can be created by chlorination of ciprofloxacin