Learn More
Proteochem Inc DSG Crosslinker, 79642-50-5, MFCD00153597, 100 mg
Supplier:  Proteochem Inc C1104100MG
Encompass_Preferred
MF: C13 H14 N2 O8, MW: 326.26, Alternative names: Di(N-succinimidyl) glutarate; Di-succinimidyl glutarate; Di N-succinimidyl glutarate; DSG Cross-linker; Bis-NHS glutarate; Disuccinimidyl glutaric dicarboxylate. DSG crosslinker is a homobifunctional crosslinking reagent that is membrane permeable. DSG protein crosslinker is often used in ChIP assays.
Catalog No. NC0054325