Learn More
Medchemexpress LLC DSG Crosslinker 100mg | 79642-50-5 | 100 MG
SDP
Catalog No. 5000229034
Encompass_Preferred
Disuccinimidyl glutarate is a homobifunctional N-hydroxysuccinimide (NHS) ester crosslinker that couples primary amines via a glutarate spacer. It is used in antibody-drug conjugate synthesis, protein and peptide crosslinking, and surface or bead immobilization in research applications. The compound is supplied at high purity for laboratory use.
  • Amine-reactive NHS ester groups enable efficient coupling of primary amines.
  • Homobifunctional design produces stable amine-to-amine crosslinks.
  • Glutarate spacer provides controlled linker length for crosslinking studies.
  • Available in small to multi-gram pack sizes to support method development.
  • Supplied for research use only; not for human or clinical use.
Catalog No. 50-002-29034 Supplier Medchemexpress LLC Supplier No. HY114697100MG
Disuccinimidyl glutarate is a homobifunctional N-hydroxysuccinimide (NHS) ester crosslinker that couples primary amines via a glutarate spacer. It is used in antibody-drug conjugate synthesis, protein and peptide crosslinking, and surface or bead immobilization in research applications. The compound is supplied at high purity for laboratory use.
  • Amine-reactive NHS ester groups enable efficient coupling of primary amines.
  • Homobifunctional design produces stable amine-to-amine crosslinks.
  • Glutarate spacer provides controlled linker length for crosslinking studies.
  • Available in small to multi-gram pack sizes to support method development.
  • Supplied for research use only; not for human or clinical use.