Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DSG Crosslinker 10mg | 79642-50-5 | 10 MG
SDP

Catalog No. 5000226368
Encompass_Preferred

DSG Crosslinker (disuccinimidyl glutarate) is a homobifunctional N-hydroxysuccinimide (NHS) ester crosslinker used as a cleavable ADC linker for antibody-drug conjugation and for amine-to-amine coupling in bioconjugation workflows.

  • Homobifunctional NHS ester for amine coupling.
  • Molecular weight 326.26 g/mol and formula C13H14N2O8.
  • High purity (98.0%) suitable for research use.
  • Soluble in DMSO (100 mg/mL) with ultrasonic assistance.
  • Powder stable at -20°C for extended storage.

Catalog No. 50-002-26368 Supplier Medchemexpress LLC Supplier No. HY11469710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DSG Crosslinker (disuccinimidyl glutarate) is a homobifunctional N-hydroxysuccinimide (NHS) ester crosslinker used as a cleavable ADC linker for antibody-drug conjugation and for amine-to-amine coupling in bioconjugation workflows.

  • Homobifunctional NHS ester for amine coupling.
  • Molecular weight 326.26 g/mol and formula C13H14N2O8.
  • High purity (98.0%) suitable for research use.
  • Soluble in DMSO (100 mg/mL) with ultrasonic assistance.
  • Powder stable at -20°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.