Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proteochem Inc DSG CROSSLINKER 1GM

Catalog No. NC1117385
Encompass_Preferred

  • Homobifunctional crosslinker for protein modification.
  • Membrane-permeable for enhanced cell interaction.
  • Amines-reactive NHS esters form stable amide bonds.
  • 5-atom spacer arm measuring 7.7 Å for flexibility in binding.
  • Chemical formula: C13 H14 N2 O8.
  • Molecular weight: 326.26 g/mol.
  • CAS Number: 79642-50-5.
  • Purity ≥ 97% as determined by quantitative NMR.
  • Solubility in DMSO or DMF is required.
  • Effective in ChIP assays and Region Capture Micro-C protocols.
  • Optimal reaction pH range of 7 to 9 for primary amines.

Catalog No. NC1117385 Supplier Proteochem Inc Supplier No. C11041GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Homobifunctional crosslinker for protein modification.
  • Membrane-permeable for enhanced cell interaction.
  • Amines-reactive NHS esters form stable amide bonds.
  • 5-atom spacer arm measuring 7.7 Å for flexibility in binding.
  • Chemical formula: C13 H14 N2 O8.
  • Molecular weight: 326.26 g/mol.
  • CAS Number: 79642-50-5.
  • Purity ≥ 97% as determined by quantitative NMR.
  • Solubility in DMSO or DMF is required.
  • Effective in ChIP assays and Region Capture Micro-C protocols.
  • Optimal reaction pH range of 7 to 9 for primary amines.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.