Learn More
Medchemexpress LLC DSG Crosslinker 50mg | 79642-50-5 | 50 MG
SDP
Catalog No. 5000229608
Encompass_Preferred
Disuccinimidyl glutarate (DSG) is a homobifunctional N-hydroxysuccinimide (NHS) ester crosslinker used to conjugate primary amines in ADC synthesis and general bioconjugation workflows. It is supplied as a solid reagent for laboratory-scale crosslinking and protein modification.
  • Reacts with primary amines to form stable amide bonds.
  • Applicable to antibody-drug conjugate linker synthesis and peptide/protein crosslinking.
  • Available in small, convenient pack sizes for research use.
  • Molecular weight 326.26 g/mol and formula C13H14N2O8.
Catalog No. 50-002-29608 Supplier Medchemexpress LLC Supplier No. HY11469750MG
Disuccinimidyl glutarate (DSG) is a homobifunctional N-hydroxysuccinimide (NHS) ester crosslinker used to conjugate primary amines in ADC synthesis and general bioconjugation workflows. It is supplied as a solid reagent for laboratory-scale crosslinking and protein modification.
  • Reacts with primary amines to form stable amide bonds.
  • Applicable to antibody-drug conjugate linker synthesis and peptide/protein crosslinking.
  • Available in small, convenient pack sizes for research use.
  • Molecular weight 326.26 g/mol and formula C13H14N2O8.