Learn More
Medchemexpress LLC DSG Crosslinker 5mg | 79642-50-5 | 326.26 g·mol^-1 | C13H14N2O8 | 5 MG
SDP
Catalog No. 5000228213
Encompass_Preferred
DSG Crosslinker (disuccinimidyl glutarate) is a homobifunctional N-hydroxysuccinimide (NHS) ester crosslinker that couples primary amines via a glutarate spacer. It is used for protein crosslinking, bioconjugation, surface immobilization, and as a linker in antibody-drug conjugate (ADC) research. Supplied for research use only.
  • Reacts with primary amines to form stable amide bonds.
  • Homobifunctional NHS-ester enabling amine-to-amine crosslinking.
  • Membrane-permeable, suitable for intracellular crosslinking.
  • Applicable to ADC synthesis, protein conjugation, and surface immobilization.
  • Typically supplied at ~98.0% purity in small research pack sizes.
Catalog No. 50-002-28213 Supplier Medchemexpress LLC Supplier No. HY1146975MG
DSG Crosslinker (disuccinimidyl glutarate) is a homobifunctional N-hydroxysuccinimide (NHS) ester crosslinker that couples primary amines via a glutarate spacer. It is used for protein crosslinking, bioconjugation, surface immobilization, and as a linker in antibody-drug conjugate (ADC) research. Supplied for research use only.
  • Reacts with primary amines to form stable amide bonds.
  • Homobifunctional NHS-ester enabling amine-to-amine crosslinking.
  • Membrane-permeable, suitable for intracellular crosslinking.
  • Applicable to ADC synthesis, protein conjugation, and surface immobilization.
  • Typically supplied at ~98.0% purity in small research pack sizes.