Learn More
Cayman Chemical DSG-PEG 2000 1g
SDP
Catalog No. 502916819
Encompass_Preferred
A PEGylated derivative of DSG; LNPs containing DSG-mPEG(2000) and encapsulating an imidazoquinoline TLR7 and TLR8 agonist and a peptide antigen EAch conjugated to PGA increase the activation of dendritic cells, macrophages, and B cells in the draining lymph node and dendritic cells and B cells in the spleen in mice compared to the non-LNP-encapsulated PGA-conjugated TLR7/8 agonist and peptide antigen.
Catalog No. 50-291-6819 Supplier Cayman Chemical Supplier No. 378001 G
A PEGylated derivative of DSG; LNPs containing DSG-mPEG(2000) and encapsulating an imidazoquinoline TLR7 and TLR8 agonist and a peptide antigen EAch conjugated to PGA increase the activation of dendritic cells, macrophages, and B cells in the draining lymph node and dendritic cells and B cells in the spleen in mice compared to the non-LNP-encapsulated PGA-conjugated TLR7/8 agonist and peptide antigen.