Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Avanti Polar Lipids DSG-PEG 2000 | 308805-39-2 | 1 g

Catalog No. NC1604841
Encompass

DSG-PEG 2000 | Purity: 99% | Mol Wt: 2619.760 | 308805-39-2 | 1 g

Catalog No. NC1604841 Supplier Avanti Polar Lipids Supplier No. 880152P1G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DSG-PEG 2000 | Purity: 99% | Mol Wt: 2619.760 | 308805-39-2 | 1 g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.