Learn More
Medchemexpress LLC Dsg-peg | 308805-39-2 | MFCD31813707 | 100.0% | 2000 (average) | (C2H4O)nC40H78O5 | 500 MG
SDP
Catalog No. 5000284571
Encompass_Preferred
DSG-PEG (MW 2000) is a polyethylene glycol-conjugated lipid supplied as a powder and used to prepare liposomes and other lipid-based delivery systems. It provides PEGylation to improve particle stability and circulation, and is available in multiple package sizes.
  • Pegylated lipid for liposome and nanoparticle formulation
  • Average molecular weight 2000
  • High purity (reported 99.97%)
  • Powder form with recommended refrigerated or frozen storage for extended stability
  • Available in multiple package sizes, including 500 MG
Catalog No. 50-002-84571 Supplier Medchemexpress LLC Supplier No. HYW591381500MG
DSG-PEG (MW 2000) is a polyethylene glycol-conjugated lipid supplied as a powder and used to prepare liposomes and other lipid-based delivery systems. It provides PEGylation to improve particle stability and circulation, and is available in multiple package sizes.
  • Pegylated lipid for liposome and nanoparticle formulation
  • Average molecular weight 2000
  • High purity (reported 99.97%)
  • Powder form with recommended refrigerated or frozen storage for extended stability
  • Available in multiple package sizes, including 500 MG