Learn More
Medchemexpress LLC Dsg-PEG (MW 2000) | 308805-39-2 | MFCD31813707 | 100.0% | 2000 (average) | C42H82O6 | 250 MG
SDP
Catalog No. 5000284024
Encompass_Preferred
DSG-PEG (MW 2000) is a PEGylated lipid used as a steric-stabilizing component in liposome and lipid nanoparticle formulations for research. It provides a methoxy-terminated polyethylene glycol chain (average MW 2000) conjugated to a distearoyl glycerol backbone, supplied as a high-purity reagent for formulation studies.
  • Pegylated lipid with average molecular weight 2000.
  • Provides steric stabilization to liposomes and lipid nanoparticles.
  • Suitable for liposome preparation and PEGylated formulations.
  • High purity suitable for research applications.
  • Available in milligram and gram pack sizes for formulation work.
Catalog No. 50-002-84024 Supplier Medchemexpress LLC Supplier No. HYW591381250MG
DSG-PEG (MW 2000) is a PEGylated lipid used as a steric-stabilizing component in liposome and lipid nanoparticle formulations for research. It provides a methoxy-terminated polyethylene glycol chain (average MW 2000) conjugated to a distearoyl glycerol backbone, supplied as a high-purity reagent for formulation studies.
  • Pegylated lipid with average molecular weight 2000.
  • Provides steric stabilization to liposomes and lipid nanoparticles.
  • Suitable for liposome preparation and PEGylated formulations.
  • High purity suitable for research applications.
  • Available in milligram and gram pack sizes for formulation work.