Learn More
Apexbio Technology LLC DSG-PEG 2000 100mg
SDP
Catalog No. 503107310
Encompass_Preferred
DSG-PEG 2000 is a PEG-modified lipid that can be used with cationic lipids such as D-Lin-MC3-DMA (No A8791) to prepare cationic nanoparticles (LNP) capable of encapsulating and delivering siRNA mRNA DNA or small molecules into the cytoplasm
Catalog No. 50-310-7310 Supplier Apexbio Technology LLC Supplier No. M2002100
DSG-PEG 2000 is a PEG-modified lipid that can be used with cationic lipids such as D-Lin-MC3-DMA (No A8791) to prepare cationic nanoparticles (LNP) capable of encapsulating and delivering siRNA mRNA DNA or small molecules into the cytoplasm