Learn More
Aviva Systems Biology Corp DSG2 Antibody - N-terminal region (ARP45198_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45198_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DSG2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DSG2. Alternate names: ARVC10, ARVD10, CDHF5, HDGC, MGC117034, MGC117036, MGC117037, CMD1BBApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58981