Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 1830MSM5P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: DSG3/1535. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: A431 cells. Skin or Lung squamous cell carcinoma. Cellular Staining: Cell surface. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human DSG3 extracellular protein fragment. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 130kDa.Chromosome: 18q12.1. Gene Symbol: DSG3. Gene ID: 1830. Ensemble: ENSG00000134757. Uniprot: NP_001935.
Catalog No. 50-188-4653