Learn More
Aviva Systems Biology Corp DSG3 Antibody - C-terminal region (ARP61265_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61265_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DSG3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human DSG3. Alternate names: CDHF6, DKFZp686P23184, PVAApplicationWB. Species reactivity: Human
Catalog No. 50-287-317