Learn More
Lumiprobe dsGreen Gel Staining Solution, 1 mL
Catalog No. 501143041
Encompass_Preferred
dsGreen, an equivalent of SYBR Green I is a sensitive dsDNA binding dye, which can be used for routine DNA detection in agarose and polyacrylamide gels.
Catalog No. 50-114-3041 Supplier Lumiprobe Supplier No. 20010
dsGreen, an equivalent of SYBR Green I is a sensitive dsDNA binding dye, which can be used for routine DNA detection in agarose and polyacrylamide gels.