Learn More
Andwin Scientific DSH WIEGHING ALUM 50ML 1000/CS
SDP
Catalog No. NC9221298
Encompass_Preferred
The aluminum crinkle dishes are supreme products for weighing, dispensing, and storing purposes. The dishes can also be utilized as vaporating dishes, parts containers, or dust covers.This product is specially designed with tabs that allow easy and convenient handling and crimped sides for sturdiness. This product has a capacity of 60 mL or 2 oz and is sold in a case of 1440.
Catalog No. NC9221298 Supplier Andwin Scientific Supplier No. 25433008
The aluminum crinkle dishes are supreme products for weighing, dispensing, and storing purposes. The dishes can also be utilized as vaporating dishes, parts containers, or dust covers.This product is specially designed with tabs that allow easy and convenient handling and crimped sides for sturdiness. This product has a capacity of 60 mL or 2 oz and is sold in a case of 1440.