Learn More
Medchemexpress LLC DSHS00884 | 675104-49-1 | 99.8% | 308.38 | 10 MG
SDP
Catalog No. 5000436383
Encompass_Preferred
DSHS00884 is a potent human papillomavirus E6 inhibitor. It stabilizes p53 levels in HPV-positive cells by blocking E6-mediated p53 degradation.
  • Potent human papillomavirus E6 inhibitor.
  • Stabilizes p53 levels in HPV-positive cells.
  • Blocks E6-mediated p53 degradation.
  • Soluble in DMSO.
  • Can be formulated for in vivo studies.
Catalog No. 50-004-36383 Supplier Medchemexpress LLC Supplier No. HY11379410MG
DSHS00884 is a potent human papillomavirus E6 inhibitor. It stabilizes p53 levels in HPV-positive cells by blocking E6-mediated p53 degradation.
  • Potent human papillomavirus E6 inhibitor.
  • Stabilizes p53 levels in HPV-positive cells.
  • Blocks E6-mediated p53 degradation.
  • Soluble in DMSO.
  • Can be formulated for in vivo studies.