Learn More
Vector Laboratories Datura Stramonium Lectin (DSL), Unconjugated, 5ml
SDP
Supplier:  Vector Laboratories L11805
Encompass
The carbohydrate binding site recognizes (β-1,4) linked N-acetylglucosamine oligomers, preferring chitobiose or chitotriose over a single N-acetylglucosamine residue. This lectin binds well in the acidic pH range but its affinity decreases above pH 8.0. DSL also binds well to N-acetyllactosamine and oligomers containing repeating N-acetyllactosamine sequences. A branched pentasaccharide including two N-acetyllactosamine disaccharides linked to mannose (β-1,6) and (β-1,2) was reported to be the most potent inhibitor of agglutination.
Catalog No. 50-204-5672