Learn More
Medchemexpress LLC DSM705 hydrochloride | 2989908-08-7 | 99.6% | C19H20ClF3N6O | 10MG
SDP
Catalog No. 5000260878
Encompass_Preferred
DSM705 hydrochloride is the hydrochloride salt of DSM705, a pyrrole-based dihydroorotate dehydrogenase (DHODH) inhibitor with oral antimalarial activity. It shows nanomolar potency against Plasmodium DHODH (IC50 ≈95 nM for P. falciparum and ≈52 nM for P. vivax) and activity against Plasmodium parasites, with minimal inhibition of mammalian DHODHs.
  • Pyrrole-based DHODH inhibitor with nanomolar potency.
  • Orally active antimalarial compound.
  • Selective for Plasmodium DHODH over mammalian DHODHs.
  • Hydrochloride salt form for improved solubility and handling.
  • Available in multiple solid and solution pack sizes for research use.
  • Reported purity approximately 99.6%.
Catalog No. 50-002-60878 Supplier Medchemexpress LLC Supplier No. HY132171A10MG
DSM705 hydrochloride is the hydrochloride salt of DSM705, a pyrrole-based dihydroorotate dehydrogenase (DHODH) inhibitor with oral antimalarial activity. It shows nanomolar potency against Plasmodium DHODH (IC50 ≈95 nM for P. falciparum and ≈52 nM for P. vivax) and activity against Plasmodium parasites, with minimal inhibition of mammalian DHODHs.
  • Pyrrole-based DHODH inhibitor with nanomolar potency.
  • Orally active antimalarial compound.
  • Selective for Plasmodium DHODH over mammalian DHODHs.
  • Hydrochloride salt form for improved solubility and handling.
  • Available in multiple solid and solution pack sizes for research use.
  • Reported purity approximately 99.6%.