Learn More
Medchemexpress LLC DSM705 hydrochloride | 2989908-08-7 | MFCD34676639 | 99.6% | 440.85 g/mol | C19H20ClF3N6O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY132171A5MG
Encompass_Preferred
DSM705 hydrochloride is a pyrrole-based dihydroorotate dehydrogenase (DHODH) inhibitor developed as an orally active antimalarial compound. It shows nanomolar potency against Plasmodium DHODHs and parasites while exhibiting no inhibition of mammalian DHODHs. The substance is supplied as a hydrochloride salt, white to off-white solid, with molecular weight 440.85 g/mol and typical purity ~99.56%.
  • Potent DHODH inhibitor with nanomolar activity against Plasmodium enzymes.
  • Selective for parasite DHODH over mammalian DHODHs, reducing off-target effects.
  • Suitable for in vitro and in vivo antimalarial research applications.
  • High purity enables reliable biochemical and cellular assays.
  • Available in small research quantities for early-stage studies.
Catalog No. 50-002-62505