Learn More
Cayman Chemical DSmethyl MIrtazapInhydroc 10mg
SDP
Catalog No. 502949199
Encompass_Preferred
A metabolite of mirtazapine; formed from mirtazapine by CYP3A4
Catalog No. 50-294-9199 Supplier Cayman Chemical Supplier No. 3426110 MG
A metabolite of mirtazapine; formed from mirtazapine by CYP3A4