Learn More
Aviva Systems Biology Corp DSN1 Antibody - C-terminal region (ARP65750_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65750_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DSN1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human DSN1. Alternate names: C20orf172, FLJ13346, KNL3, MGC32987, MIS13, dJ469A13.2, hKNL-3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-560