Learn More
Metrohm USA Digital combined pH electrode, for non-aqueous titrations, glass membrane, flexible ground-joint diaphragm, 0-14 pH, 0-70 °C, 12 mm diameter, Metrohm plug-in head Q
SDP
Supplier:  Metrohm USA 600203300
Encompass
The Metrohm dSolvotrode is a digital, combined pH electrode designed for accurate pH measurements in non-aqueous acidic and basic titrations. This electrode features a specially developed glass membrane optimized for poorly conducting solutions, providing reliable performance across challenging environments. It comes equipped with a flexible ground-joint diaphragm that enhances its usability with contaminated samples, preventing blockage and preserving measurement accuracy. The dSolvotrode is designed for seamless operation with Metrohm OMNIS Titrators, supporting a range of non-aqueous reference electrolytes.
Catalog No. NC2239701