Learn More
AdipoGen DSP-1
Supplier: AdipoGen CDXD0391M001
Chemical. CAS 1609395-29-0. Formula C28H16FNO8. MW 494.44. Synthetic Fluorescent probe for detection of H2S2 and hydrogen polysulfides in aqueous buffers.