Learn More
Medchemexpress LLC HY-103210 10mg Medchemexpress, DSP-4 (hydrochloride) CAS:40616-75-9 Purity:>98%
SDP
Catalog No. 501968056
Encompass_Preferred
Medchemexpress, HY-103210 10mg DSP-4 (HCl) CAS:40616-75-9 DSP-4 hydrochloride (Neurotoxin DSP 4 hydrochloride) is a highly selective neurotoxin and readily passes the blood-brain barrier with neurotoxic effects on noradrenergic neurons of adult and developing rats, can be used for the temporary selective degradation of the central and peripheral noradrenergic neurons, mainly those from the locus coeruleus (LC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8056 Supplier Medchemexpress LLC Supplier No. HY10321010MG
Medchemexpress, HY-103210 10mg DSP-4 (HCl) CAS:40616-75-9 DSP-4 hydrochloride (Neurotoxin DSP 4 hydrochloride) is a highly selective neurotoxin and readily passes the blood-brain barrier with neurotoxic effects on noradrenergic neurons of adult and developing rats, can be used for the temporary selective degradation of the central and peripheral noradrenergic neurons, mainly those from the locus coeruleus (LC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.