Learn More
Medchemexpress LLC DSP Crosslinker | 57757-57-0 | 98.04% | 404.42 | 250 MG
SDP
Catalog No. 5000439716
Encompass_Preferred
DSP Crosslinker is an amine-reactive and cleavable protein crosslinker. It enables stable protein-protein, peptide, and biomolecule conjugation.
  • Amine-reactive and cleavable protein crosslinker
  • Enables stable protein-protein, peptide, and biomolecule conjugation
Catalog No. 50-004-39716 Supplier Medchemexpress LLC Supplier No. 250MGHY118759
Add to Cart
Add to Cart
DSP Crosslinker is an amine-reactive and cleavable protein crosslinker. It enables stable protein-protein, peptide, and biomolecule conjugation.
  • Amine-reactive and cleavable protein crosslinker
  • Enables stable protein-protein, peptide, and biomolecule conjugation