Learn More
Electron Microscopy Sciences EMS Disposable Scissors, Straight Blades Sharp/Blunt, 4-1/2"
SDP
Catalog No. 501931454
Encompass_Preferred
Disposable scissors with a straight cutting edge, these scissors are highly reommended for infectious cases. Disposable instruments designed specifically for single use only. They are not autoclavable.
Blade: Straight Sharp/Blunt
OAL: 4-1/2"
Catalog No. 50-193-1454 Supplier Electron Microscopy Sciences Supplier No. 7299646
Disposable scissors with a straight cutting edge, these scissors are highly reommended for infectious cases. Disposable instruments designed specifically for single use only. They are not autoclavable.
Blade: Straight Sharp/Blunt
OAL: 4-1/2"