Learn More
Electron Microscopy Sciences Disposable Base Mold for Macrocassettes
SDP
Catalog No. 5030334
Encompass_Preferred
Made from PVC with excellent thermal exchange. Specially designed for the Macro-Cassette above. It has a smooth interior finish and rounded corners to facilitate its removal. Will resist temperatures from –80 degree C to 60 degree C. Not to be used with xylene or acetone. Measures: 37 x 24 x 10 mm, and 5 ml volume.
Catalog No. 50-303-34 Supplier Electron Microscopy Sciences Supplier No. 62353
Made from PVC with excellent thermal exchange. Specially designed for the Macro-Cassette above. It has a smooth interior finish and rounded corners to facilitate its removal. Will resist temperatures from –80 degree C to 60 degree C. Not to be used with xylene or acetone. Measures: 37 x 24 x 10 mm, and 5 ml volume.