Learn More
Electron Microscopy Sciences Disposable Blade Knife Handle #6, 5 3/16"
SDP
Catalog No. 50949321
Encompass_Preferred
These strong, molded plastic handles provide a secure palm-fitting grip for postmortem work, trimming light plaster casts, and similar forceful cutting.
OAL: 5 3/16"
Use: Size 20, 21, 22, 23 blades
Catalog No. 50-949-321 Supplier Electron Microscopy Sciences Supplier No. 7204006
These strong, molded plastic handles provide a secure palm-fitting grip for postmortem work, trimming light plaster casts, and similar forceful cutting.
OAL: 5 3/16"
Use: Size 20, 21, 22, 23 blades