Learn More
Electron Microscopy Sciences Disposable Blade Knife Handle #8, 5 9/32"
SDP
Catalog No. 50949336
Encompass_Preferred
These strong, molded plastic handles provide a secure palm-fitting grip for postmortem work, trimming light plaster casts, and similar forceful cutting.
OAL: 5 9/32"
Use: Size 60 blades
Catalog No. 50-949-336 Supplier Electron Microscopy Sciences Supplier No. 7204008
These strong, molded plastic handles provide a secure palm-fitting grip for postmortem work, trimming light plaster casts, and similar forceful cutting.
OAL: 5 9/32"
Use: Size 60 blades