Learn More
Electron Microscopy Sciences Disposable Blade Knife Handles #5, 4-3/16"
SDP
Catalog No. 50949320
Encompass_Preferred
These strong, molded plastic handles provide a secure palm-fitting grip for postmortem work, trimming light plaster casts, and similar forceful cutting.
OAL: 4-3/16"
Use: Size 10, 11, 12, 15 blades
Catalog No. 50-949-320 Supplier Electron Microscopy Sciences Supplier No. 7204005
These strong, molded plastic handles provide a secure palm-fitting grip for postmortem work, trimming light plaster casts, and similar forceful cutting.
OAL: 4-3/16"
Use: Size 10, 11, 12, 15 blades