Learn More
Electron Microscopy Sciences Disposable Microtome Blades Premium, 3"L x 0.498"W, 0.02" thick
SDP
Catalog No. 5025531
Encompass_Preferred
These stainless steel disposable blades feature an edge that is the product of years of research. Repeated testing was performed by histologists to assure that SHUR/SHARP blades represent the ultimate in consistency for sectioning technology. Chatterless and compression free ribbons will become routine no matter what the tissue is. Heavy duty disposable premium microtome blades, Uncoated.
Size: 3"L x 0.498"W
Thickness: 0.02"
Catalog No. 50-255-31 Supplier Electron Microscopy Sciences Supplier No. 6306101
These stainless steel disposable blades feature an edge that is the product of years of research. Repeated testing was performed by histologists to assure that SHUR/SHARP blades represent the ultimate in consistency for sectioning technology. Chatterless and compression free ribbons will become routine no matter what the tissue is. Heavy duty disposable premium microtome blades, Uncoated.
Size: 3"L x 0.498"W
Thickness: 0.02"