Learn More
Echelon Biosciences Research Labs DSPC 250 mg
SDP
Supplier:  Echelon Biosciences Research Labs L1118250MG
Encompass
1,2-Distearoyl-sn-glycero-3-phosphocholine (DSPC) is a phosphatidylcholine with stearoyl (octadecanoyl) acyl groups. It derives from an octadecanoic acid.Phosphatidylcholine (PC) is generally the most abundant lipid in animal cell membranes providing structural framework. PC is more common in the outer leaflet where it functions as part of the permeability barrier. PC is also the primary substrate of Phospholipase D enzymes which produce the signaling lipids phosphatidic acid and lysophosphatidic acid. This item is non returnable
Catalog No. 50-720-492