Learn More
Apexbio Technology LLC 1 2-Distearoyl-sn-glycero-3-PC 500mg
SDP
Catalog No. 503107439
Encompass_Preferred
1 2-Distearoyl-sn-glycero-3-phosphorylcholine (DSPC) is a phospholipid used in the synthesis of liposomes and lipid nanoparticle (LNP) systems for research related to mRNA vaccines or drug delivery systems
Catalog No. 50-310-7439 Supplier Apexbio Technology LLC Supplier No. C4911250
1 2-Distearoyl-sn-glycero-3-phosphorylcholine (DSPC) is a phospholipid used in the synthesis of liposomes and lipid nanoparticle (LNP) systems for research related to mRNA vaccines or drug delivery systems