Learn More
Apexbio Technology LLC 1 2-Distearoyl-sn-glycero-3-PE 250mg
SDP
Catalog No. 503107126
Encompass_Preferred
1 2-Distearoyl-sn-glycero-3-phosphorylethanolamine (DSPE) is a phosphatidylethanolamine (PE) lipid that can be used for preparing liposomes employed in drug delivery gene transfection and vaccine delivery
Catalog No. 50-310-7126 Supplier Apexbio Technology LLC Supplier No. C4855250
1 2-Distearoyl-sn-glycero-3-phosphorylethanolamine (DSPE) is a phosphatidylethanolamine (PE) lipid that can be used for preparing liposomes employed in drug delivery gene transfection and vaccine delivery