Learn More
Medchemexpress LLC DSPE-Biotin | 133695-76-8 | 97.5% | 974.36 g·mol⁻¹ | C51H96N3O10PS | 100 MG
SDP
Catalog No. 5000280976
Encompass_Preferred
DSPE-Biotin is a biotin-functionalized phospholipid used to introduce biotin groups into lipid membranes and liposome surfaces for biochemical and bioconjugation applications. It enables high-affinity capture and labeling of lipid particles via streptavidin/avidin interactions and is supplied for research use with recommended storage protected from light at low temperature.
  • Biotin-functionalized phospholipid for membrane and liposome functionalization.
  • Enables high-affinity binding to streptavidin/avidin systems for capture and targeting.
  • Suitable for bioconjugation, labeling, and assay development with lipid particles.
  • High reported purity (97.5%).
  • Supplied as 100 MG and intended for research use only.
  • Store protected from light at -20 °C for long-term stability.
Catalog No. 50-002-80976 Supplier Medchemexpress LLC Supplier No. HYW441000100MG
DSPE-Biotin is a biotin-functionalized phospholipid used to introduce biotin groups into lipid membranes and liposome surfaces for biochemical and bioconjugation applications. It enables high-affinity capture and labeling of lipid particles via streptavidin/avidin interactions and is supplied for research use with recommended storage protected from light at low temperature.
  • Biotin-functionalized phospholipid for membrane and liposome functionalization.
  • Enables high-affinity binding to streptavidin/avidin systems for capture and targeting.
  • Suitable for bioconjugation, labeling, and assay development with lipid particles.
  • High reported purity (97.5%).
  • Supplied as 100 MG and intended for research use only.
  • Store protected from light at -20 °C for long-term stability.