Learn More
Medchemexpress LLC Dspe-biotin | 133695-76-8 | 97.5% | 974.36 g/mol | C51H96N3O10PS | 5 MG
SDP
Catalog No. 5000281011
Encompass_Preferred
DSPE-Biotin is a biotinylated phospholipid designed to introduce biotin functional groups into liposomes, supported lipid bilayers, and other lipid-based systems for labeling, affinity capture, and biochemical assays. It is provided at high purity for research applications and should be handled and stored according to the supplier's recommendations.
  • Biotinylated lipid for affinity capture and labeling.
  • Compatible with liposome and supported bilayer formulations.
  • High purity for reproducible experimental results.
  • Soluble in DMSO (≈1.3 mg/mL) with ultrasonic warming to aid dissolution.
  • Recommended long-term storage at -20°C protected from light.
  • Useful for biochemical assays, surface functionalization, and probe attachment.
Catalog No. 50-002-81011 Supplier Medchemexpress LLC Supplier No. HYW4410005MG
DSPE-Biotin is a biotinylated phospholipid designed to introduce biotin functional groups into liposomes, supported lipid bilayers, and other lipid-based systems for labeling, affinity capture, and biochemical assays. It is provided at high purity for research applications and should be handled and stored according to the supplier's recommendations.
  • Biotinylated lipid for affinity capture and labeling.
  • Compatible with liposome and supported bilayer formulations.
  • High purity for reproducible experimental results.
  • Soluble in DMSO (≈1.3 mg/mL) with ultrasonic warming to aid dissolution.
  • Recommended long-term storage at -20°C protected from light.
  • Useful for biochemical assays, surface functionalization, and probe attachment.