Learn More
Medchemexpress LLC DSPE-biotin | 133695-76-8 | MFCD34561787 | 97.5% | 974.36 | C51H96N3O10PS | 10 MG
SDP
Catalog No. 5000281756
Encompass_Preferred
DSPE-Biotin is a biotinylated phospholipid used to introduce biotin functionality into liposomes, lipid bilayers, and other lipid-based formulations for biochemical research and bioconjugation. It facilitates specific capture, labeling, and immobilization through high-affinity streptavidin/avidin interactions.
  • Provides biotin functionality for liposomes and lipid bilayers.
  • Enables high-affinity binding to streptavidin and avidin.
  • Suitable for liposome formulation and surface immobilization applications.
  • Soluble in DMSO; ultrasonic warming improves solubility.
  • High purity for research use; handle and store protected from light.
  • Stable when stored at -20°C for long-term storage.
Catalog No. 50-002-81756 Supplier Medchemexpress LLC Supplier No. HYW44100010MG
DSPE-Biotin is a biotinylated phospholipid used to introduce biotin functionality into liposomes, lipid bilayers, and other lipid-based formulations for biochemical research and bioconjugation. It facilitates specific capture, labeling, and immobilization through high-affinity streptavidin/avidin interactions.
  • Provides biotin functionality for liposomes and lipid bilayers.
  • Enables high-affinity binding to streptavidin and avidin.
  • Suitable for liposome formulation and surface immobilization applications.
  • Soluble in DMSO; ultrasonic warming improves solubility.
  • High purity for research use; handle and store protected from light.
  • Stable when stored at -20°C for long-term storage.