Learn More
Medchemexpress LLC Dspe-m-peg-nhs | 99.4% | 3,400 (average) | 50 MG
SDP
Catalog No. 5000281111
Encompass_Preferred
DSPE-m-PEG-NHS (MW 3400) is a PEGylated phospholipid bearing an N-hydroxysuccinimide (NHS) ester. Supplied as a white-to-off-white solid, it is used to formulate liposomes and lipid nanoparticles and enables amine-reactive surface conjugation for targeted delivery applications such as DNA and mRNA vaccines.
  • Provides an NHS-reactive ester for efficient conjugation to primary amines.
  • Facilitates PEGylation to improve nanoparticle stability and circulation time.
  • Suitable for liposome and lipid nanoparticle formulation.
  • Average molecular weight about 3,400 for consistent performance.
  • Stable as a powder when stored at -20°C, supporting extended shelf life.
Catalog No. 50-002-81111 Supplier Medchemexpress LLC Supplier No. HYW441015A50MG
DSPE-m-PEG-NHS (MW 3400) is a PEGylated phospholipid bearing an N-hydroxysuccinimide (NHS) ester. Supplied as a white-to-off-white solid, it is used to formulate liposomes and lipid nanoparticles and enables amine-reactive surface conjugation for targeted delivery applications such as DNA and mRNA vaccines.
  • Provides an NHS-reactive ester for efficient conjugation to primary amines.
  • Facilitates PEGylation to improve nanoparticle stability and circulation time.
  • Suitable for liposome and lipid nanoparticle formulation.
  • Average molecular weight about 3,400 for consistent performance.
  • Stable as a powder when stored at -20°C, supporting extended shelf life.