Learn More
Medchemexpress LLC Dspe-m-peg-nhs | 99.4% | 3,400 Da (average) | 25 MG
SDP
Catalog No. 5000281017
Encompass_Preferred
DSPE-m-PEG-NHS (MW 3400) is a PEGylated phospholipid with an N-hydroxysuccinimide (NHS) activated ester on the PEG terminus. Supplied as a powder with an average molecular weight of approximately 3,400 Da, it is used to introduce PEG-lipid functionality into liposomes and lipid nanoparticles and to enable covalent attachment of amine-containing ligands for targeted delivery and surface modification.
  • NHS-activated ester for efficient amide bond formation with primary amines.
  • Average molecular weight approximately 3,400 Da for predictable PEG length.
  • Powder form suitable for storage under frozen conditions.
  • Compatible with liposome and lipid nanoparticle formulation workflows.
  • Enables surface conjugation of proteins, peptides, or antibodies.
  • High purity to support reproducible formulation and conjugation performance.
Catalog No. 50-002-81017 Supplier Medchemexpress LLC Supplier No. HYW441015A25MG
DSPE-m-PEG-NHS (MW 3400) is a PEGylated phospholipid with an N-hydroxysuccinimide (NHS) activated ester on the PEG terminus. Supplied as a powder with an average molecular weight of approximately 3,400 Da, it is used to introduce PEG-lipid functionality into liposomes and lipid nanoparticles and to enable covalent attachment of amine-containing ligands for targeted delivery and surface modification.
  • NHS-activated ester for efficient amide bond formation with primary amines.
  • Average molecular weight approximately 3,400 Da for predictable PEG length.
  • Powder form suitable for storage under frozen conditions.
  • Compatible with liposome and lipid nanoparticle formulation workflows.
  • Enables surface conjugation of proteins, peptides, or antibodies.
  • High purity to support reproducible formulation and conjugation performance.