Learn More
Medchemexpress LLC DSPE-Mal | 1360858-99-6 | 95.0% | 899.18 g/mol | C48H87N2O11P | 25 MG
SDP
Catalog No. 5000281013
Encompass_Preferred
DSPE-Mal is a maleimide-functionalized distearoyl phospholipid used to prepare liposomes and lipid-based nanocarriers. The maleimide group enables covalent coupling to thiol-containing molecules for surface functionalization, facilitating targeted delivery and conjugation of peptides, proteins, or other ligands. Supplied as a white to off-white solid for research use.
  • Enables covalent coupling to thiol-containing molecules via maleimide-thiol chemistry.
  • Facilitates preparation of liposomes and lipid-based nanocarriers.
  • Suitable for surface functionalization with peptides, proteins, or targeting ligands.
  • Supplied as a white to off-white solid with recommended frozen storage for long-term stability.
  • Typical purity ~95.0% and molecular weight ~899.18 g/mol.
Catalog No. 50-002-81013 Supplier Medchemexpress LLC Supplier No. HYW44100725MG
DSPE-Mal is a maleimide-functionalized distearoyl phospholipid used to prepare liposomes and lipid-based nanocarriers. The maleimide group enables covalent coupling to thiol-containing molecules for surface functionalization, facilitating targeted delivery and conjugation of peptides, proteins, or other ligands. Supplied as a white to off-white solid for research use.
  • Enables covalent coupling to thiol-containing molecules via maleimide-thiol chemistry.
  • Facilitates preparation of liposomes and lipid-based nanocarriers.
  • Suitable for surface functionalization with peptides, proteins, or targeting ligands.
  • Supplied as a white to off-white solid with recommended frozen storage for long-term stability.
  • Typical purity ~95.0% and molecular weight ~899.18 g/mol.