Learn More
Medchemexpress LLC DSPE-N3 | 2839508-98-2 | 96.4% | 831.11 g·mol⁻¹ | C43H83N4O9P | 25 MG
SDP
Catalog No. 5000281010
Encompass_Preferred
DSPE-N3 is an azide-functionalized phospholipid used as a click-chemistry reagent in biochemical and nanotechnology research. It enables both copper-catalyzed and strain-promoted azide-alkyne cycloadditions for reliable conjugation of biomolecules, surface modification, and lipid nanoparticle functionalization.
  • Azide-functionalized phospholipid enabling click chemistry conjugation.
  • Compatible with CuAAC and SPAAC reaction conditions.
  • Suitable for nanoparticle and surface functionalization applications.
  • White to off-white solid with standard storage requirements.
  • Soluble in DMSO with warming and sonication for formulation.
  • High purity (~96.4%) appropriate for research use.
Catalog No. 50-002-81010 Supplier Medchemexpress LLC Supplier No. HYW44099925MG
DSPE-N3 is an azide-functionalized phospholipid used as a click-chemistry reagent in biochemical and nanotechnology research. It enables both copper-catalyzed and strain-promoted azide-alkyne cycloadditions for reliable conjugation of biomolecules, surface modification, and lipid nanoparticle functionalization.
  • Azide-functionalized phospholipid enabling click chemistry conjugation.
  • Compatible with CuAAC and SPAAC reaction conditions.
  • Suitable for nanoparticle and surface functionalization applications.
  • White to off-white solid with standard storage requirements.
  • Soluble in DMSO with warming and sonication for formulation.
  • High purity (~96.4%) appropriate for research use.