Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DSPE-N3 | 2839508-98-2 | 96.4% | 831.11 g/mol | C43H83N4O9P | 50 MG
SDP

Catalog No. 5000281503
Encompass_Preferred

DSPE-N3 is an azide-functionalized phospholipid used as a click-chemistry reagent for biochemical research. It enables covalent conjugation via azide-alkyne cycloaddition and is commonly used to functionalize liposomes, nanoparticles, and biomolecules for labeling or targeted delivery applications.

  • Contains an azide functional group for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Compatible with strain-promoted alkyne-azide cycloaddition (SPAAC) for copper-free reactions.
  • Suitable for liposome and surface functionalization in biochemical studies.
  • Supplied as a white to off-white solid with high purity.
  • Soluble in DMSO (12.5 mg/mL) with sonication and heating.
  • Recommend sealed storage at 4°C; in solution: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-81503 Supplier Medchemexpress LLC Supplier No. HYW44099950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DSPE-N3 is an azide-functionalized phospholipid used as a click-chemistry reagent for biochemical research. It enables covalent conjugation via azide-alkyne cycloaddition and is commonly used to functionalize liposomes, nanoparticles, and biomolecules for labeling or targeted delivery applications.

  • Contains an azide functional group for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Compatible with strain-promoted alkyne-azide cycloaddition (SPAAC) for copper-free reactions.
  • Suitable for liposome and surface functionalization in biochemical studies.
  • Supplied as a white to off-white solid with high purity.
  • Soluble in DMSO (12.5 mg/mL) with sonication and heating.
  • Recommend sealed storage at 4°C; in solution: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.