Learn More
Medchemexpress LLC DSPE-NHS | 1383932-86-2 | MFCD34561791 | 95.0% | 945.21 g/mol | C49H89N2O13P | 5 MG
SDP
Catalog No. 5000281717
Encompass_Preferred
DSPE-NHS is an NHS-activated phospholipid reagent used for bioconjugation and liposome/nanocarrier preparation. It contains two hydrophobic stearoyl tails and an NHS ester that reacts with primary amines to form stable amide bonds; in aqueous solution it self-assembles into lipid bilayers and is commonly used to label antibodies and conjugate proteins.
  • Reacts with primary amines to form stable amide linkages.
  • Self-assembles into lipid bilayers in aqueous solution.
  • Suitable for antibody labeling and protein conjugation.
  • Solid form with practical storage recommendations for solvent and sealed conditions.
  • Provided at high purity for research applications.
Catalog No. 50-002-81717 Supplier Medchemexpress LLC Supplier No. HYW4410115MG
DSPE-NHS is an NHS-activated phospholipid reagent used for bioconjugation and liposome/nanocarrier preparation. It contains two hydrophobic stearoyl tails and an NHS ester that reacts with primary amines to form stable amide bonds; in aqueous solution it self-assembles into lipid bilayers and is commonly used to label antibodies and conjugate proteins.
  • Reacts with primary amines to form stable amide linkages.
  • Self-assembles into lipid bilayers in aqueous solution.
  • Suitable for antibody labeling and protein conjugation.
  • Solid form with practical storage recommendations for solvent and sealed conditions.
  • Provided at high purity for research applications.