Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dspe-peg 2000 | 892144-24-0 | >99.3% | (C2H4O)nC42H82NO10P | 100 MG
SDP

Catalog No. 5000354348
Encompass_Preferred

DSPE-PEG 2000 is a PEG polymer that contains DSPE and amine end groups. It can be utilized to form micelles, which function as nanoparticles for drug delivery.

  • Application: Can be used to form micelles as nanoparticles for drug delivery.
  • Appearance: Solid, white to off-white.
  • Solubility: In DMSO: 50 mg/mL (requires ultrasonic; note that hygroscopic DMSO significantly impacts solubility, so use newly opened DMSO). In ethanol: 25 mg/mL (requires ultrasonic, warming, and heating to 60°C). In H2O: 25 mg/mL (requires ultrasonic, warming, and heating to 60°C).
  • Storage: Powder: -20°C for 3 years. In solvent: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-003-54348 Supplier Medchemexpress LLC Supplier No. HY142979100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DSPE-PEG 2000 is a PEG polymer that contains DSPE and amine end groups. It can be utilized to form micelles, which function as nanoparticles for drug delivery.

  • Application: Can be used to form micelles as nanoparticles for drug delivery.
  • Appearance: Solid, white to off-white.
  • Solubility: In DMSO: 50 mg/mL (requires ultrasonic; note that hygroscopic DMSO significantly impacts solubility, so use newly opened DMSO). In ethanol: 25 mg/mL (requires ultrasonic, warming, and heating to 60°C). In H2O: 25 mg/mL (requires ultrasonic, warming, and heating to 60°C).
  • Storage: Powder: -20°C for 3 years. In solvent: -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.