Learn More
Medchemexpress LLC DSPE-PEG 2000 | 892144-24-0 | 99.37% | (C2H4O)nC42H82NO10P | 50 MG
SDP
Catalog No. 5000353805
Encompass_Preferred
DSPE-PEG 2000 is a PEG polymer that contains DSPE and amine end groups. It can be utilized to create micelles that function as nanoparticles for drug delivery.
  • PEG polymer containing DSPE and amine end groups.
  • Can be used to form micelles.
  • Suitable for nanoparticle drug delivery.
Catalog No. 50-003-53805 Supplier Medchemexpress LLC Supplier No. HY14297950MG
DSPE-PEG 2000 is a PEG polymer that contains DSPE and amine end groups. It can be utilized to create micelles that function as nanoparticles for drug delivery.
  • PEG polymer containing DSPE and amine end groups.
  • Can be used to form micelles.
  • Suitable for nanoparticle drug delivery.