Learn More
Avanti Polar Lipids DSPE-PEG 2000 AMINE 100MG
Catalog No. NC9156675
Encompass
DSPE-PEG(2000)Amine, 1,2-Distearoyl-sn-Glycero-3-Phosphoethanolamine-N-[Amino(Polyethylene Glycol)2000] (Ammonium Salt), White Powder, Purity: >99%, Molecular Weight: 2790.52
Catalog No. NC9156675 Supplier Avanti Polar Lipids Supplier No. 880128P100MG
Add to Cart
Add to Cart
DSPE-PEG(2000)Amine, 1,2-Distearoyl-sn-Glycero-3-Phosphoethanolamine-N-[Amino(Polyethylene Glycol)2000] (Ammonium Salt), White Powder, Purity: >99%, Molecular Weight: 2790.52